A UV-Vis spectrophotometer is an analytical instrument used to measure the absorbance or transmission of ultraviolet (UV) and visible (Vis) light by a sample. It operates within the wavelength range of approximately 200 to 800 nanometers, covering both UV and visible light spectra.
Here is how it works:
The instrument uses a light source that emits both UV and visible light. The light is directed through a monochromator, which isolates specific wavelengths of light. The selected wavelength of light passes through the sample. The light that passes through the sample is then detected and measured. The instrument calculates the absorbance or transmission of the light by the sample and displays the results.
UV-Vis spectrophotometry is commonly used in various fields, including chemistry, biology, and environmental science, for applications such as:
- Determining the concentration of a substance in solution
- Characterizing the optical properties of materials
- Analyzing the kinetics of chemical reactions
- Studying the absorption spectra of compounds
The fundamental principle behind UV-Vis spectrophotometry is Beer-Lambert Law, which relates the absorbance of light to the concentration of the absorbing species in the sample:
- A=ϵ⋅c⋅l
- where:
- A is the absorbance
- ϵ is the molar absorptivity
- C is the concentration of the sample
- l is the path length of the sample cell