Robotic Pipette Tips
Pipette Specific Tips are disposable and designed to fit specific manufacturers’ single and multichannel pipettes. For best results, use the pipette tips compatible with your pipettor manufacturer. Manufacturer specific tips are available sterile or non-sterile, in a range of sizes, styles and colors and with or without filters. They may be sold in bulk packaging, in racks, individually wrapped, blister-packed or in reload formats. Universal pipette tips can be substituted for manufacturer specific tips.
Useful Links
Save Now - Exclusive Deals
Product Code 10759221
Product Code 17516696
Product Code 15378291
Product Code 12742856
Product Code 30019045
Product Code 10561972
Product Code 12716769
Product Code 30019046
Product Code 10059499
Product Code 17506696
Product Code 10623482
Product Code 12736769
Must Have
Product Code 10648221
Product Code 11751263
Product Code 17596686
Product Code 10121343
Product Code 13187444
Product Code 30019038
Product Code 10729601
Product Code 12796699
Product Code 11868423
Product Code 13427499
Product Code 15398291
Complete Your Order - Great Deals
FAQ
Robotic tips offer several benefits in automated liquid handling systems, including increased precision and consistency in liquid dispensing. They are designed to work seamlessly with robotic pipetting systems, ensuring accurate and reproducible results. Robotic tips can handle high-throughput applications efficiently, significantly speeding up workflows and reducing manual labor. They also minimize the risk of contamination and human error, which is crucial for sensitive assays and experiments. Additionally, robotic tips are available in various sizes and types, such as filter tips and low-retention tips, to meet specific experimental needs.
Proper maintenance and storage of robotic tips are essential for ensuring their performance and longevity. Store tips in a clean, dry environment to prevent contamination and damage. Keep them in their original packaging until use to maintain sterility and prevent exposure to dust and other contaminants. Regularly inspect tips for any signs of damage or defects before use. Avoid touching the tips with bare hands to prevent contamination. Ensure that the tips are compatible with the specific robotic pipetting system being used. Proper handling and storage practices will help maintain the integrity and reliability of robotic tips.
Common issues with robotic tips include improper fit or compatibility with the robotic pipetting system, which can lead to leaks or inconsistent volume dispensing. Ensure that the tips are compatible with the specific robotic system being used and are properly attached. Contamination is another issue, especially with filter tips; always handle tips with care and store them properly to maintain sterility. Clogging can occur if tips are not used correctly or if they are damaged. Regularly inspect and replace tips as needed to prevent clogging. By addressing these common issues through proper selection, handling, and storage, you can ensure the reliable performance of your robotic tips.