Fisher Scientific Logo
Produkter
Applikationer
Cellebiologi
Applikationer
Kemi
Genomik
Applikationer
Produktion og bioproces
Applikationer
Produktion og bioproces
Proteomik
Applikationer
Sikkerhed
Play Menu
Dehydrated Microbiology Media
Dehydrated Microbiology Media
Dehydrated culture media are used for the isolation, detection, and cultivation of various microorganisms. They are available in granulated or powdered forms, and in a range of formulations and formats depending on your needs.
FEATURED
Thermo Scientific™ Oxoid™ Phosphate Buffered Saline Tablets
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.
Thermo Scientific™ Oxoid™ Phosphate Buffered Saline Tablets
NON-SELECTIVE MEDIUM
Thermo Scientific™ Nutrient Agar (Dehydrated)
Cultivate and isolate fastidious pathogens
A nutritive, non-selective medium ideal for maintenance and purity check of subcultures.
Thermo Scientific™ Nutrient Agar (Dehydrated)
NUTRIENT RICH
MP Biomedicals™ Tryptic Soy Agar
Rich in nutrients for general use in microbiological laboratories
Supports the growth of a variety of microorganisms, including fastidious aerobes and anaerobes.
MP Biomedicals™ Tryptic Soy Agar
Complete Your Order - Great Deals
FAQ
Resources and Related Information
Advancing Drug Manufacturing Microbial Safety
Microbiology solutions meticulously crafted to cater to the intricate requirements of the pharmaceutical and biotechnology industries.
Advancing drug manufacturing microbial safety
100 Years of Microbiology Excellence
Microbiology product catalogue.
100 years of Microbiology Excellence
Shop By Brand