CAS RN 7558-80-7
Natriumfosfat, monobasisk, 99%, til analyse, vandfri
CAS: 7558-80-7 Molekylær formel: H2NaO4P Molekylvægt (g/mol): 119.98 MDL nummer: MFCD00003527,MFCD00146206 InChI nøgle: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 SMIL: [Na+].OP(O)([O-])=O
Natriumdihydrogenfosfat, 96%
CAS: 7558-80-7 Molekylær formel: H2NaO4P Molekylvægt (g/mol): 119.98 MDL nummer: MFCD00003527,MFCD00146206 InChI nøgle: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 IUPAC navn: natrium; dihydrogenphosphat SMIL: [Na+].OP(O)([O-])=O
Thermo Scientific Chemicals Natriumphosphat, monobasisk, 99%, til biokemi, vandfri
CAS: 7558-80-7 Molekylær formel: H2NaO4P Molekylvægt (g/mol): 119.98 MDL nummer: MFCD00003527,MFCD00146206 InChI nøgle: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 IUPAC navn: natriumdihydrogenphosphat SMIL: [Na+].OP(O)([O-])=O
Fosfatopløsning til ionkromatografi 1g/LH 2 O matrix, Fisher chemical™
CAS: 7558-80-7 Molekylvægt (g/mol): 119.98 MDL nummer: MFCD00003527,MFCD00146206 InChI nøgle: AJPJDKMHJJGVTQ-UHFFFAOYSA-M PubChem CID: 23672064 ChEBI: CHEBI:37585 SMIL: [Na+].OP(O)([O-])=O
Thermo Scientific Chemicals Fosfat, 0,5 M bufferopløsning, pH 6,5
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Fosfat, ionkromatografi standardopløsning, Specpure™ PO4∣-^3 1000 μg/ml
CAS: 7558-80-7 Molekylær formel: H2NaO4P Molekylvægt (g/mol): 119.98 MDL nummer: MFCD00003527,MFCD00146206 InChI nøgle: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: Phosphate, Ion chromatography standard solution PubChem CID: 23672064 ChEBI: CHEBI:37585 IUPAC navn: natrium; dihydrogenphosphat SMIL: [Na+].OP(O)([O-])=O
Thermo Scientific Chemicals Fosfat, 0,5 M bufferopløsning, pH 7,5
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,2 M bufferopløsning, pH 4,4
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,2 M bufferopløsning, pH 7,2
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,5 M bufferopløsning, pH 8,0
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,2 M bufferopløsning, pH 7,0
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,5 M bufferopløsning, pH 7,4
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,2 M bufferopløsning, pH 7,4
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976
Thermo Scientific Chemicals Fosfat, 0,2 M bufferopløsning, pH 7,5
CAS: 7558-80-7 Molekylvægt (g/mol): 119.976