accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 150-25-4

NaHONHOOO

CAS RN 150-25-4

IUPAC Name: sodium 2-[bis(2-hydroxyethyl)amino]acetate
Synonyms: bicine
Molecular Weight (g/mol): 185.16
Molecular Formula: C6H12NNaO4
InChi Key: MFBDBXAVPLFMNJ-UHFFFAOYSA-M
SMILES: [Na+].OCCN(CCO)CC([O-])=O
Quantity 
  • (4)
  • (6)
  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (24)
Physical Form 
  • (5)
  • (8)
  • (7)
  • (3)
pH 
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (5)
  • (3)
  • (2)
  • (3)

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (6)
  • (13)
  • (1)
  • (4)
  • (6)
  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (24)
  • (5)
  • (8)
  • (7)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.