Learn More
Uvex™ Men-T-Shirt 7459-Black 100% Cotton
Black classical T-shirt form of combed cotton, and Single Jersey with neck tape and elastic cuffs on neck
124.00 DKK
Specifications
Gender Men's
Material Cotton
Color Black
Neck Style Hooded
Seam Type Taped
View More Specs
Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17205521
View Documents
Uvex™
8816615
4X-Large
124.00 DKK
Each
Only null left
In Stock
Edge
Add to basket
17215521
View Documents
Uvex™
8816616
5X-Large
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17225521
View Documents
Uvex™
8816617
6X-Large
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17235511
View Documents
Uvex™
8816608
X-Small
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17245511
View Documents
Uvex™
8816609
Small
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17255511
View Documents
Uvex™
8816610
Medium
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17265511
View Documents
Uvex™
8816611
Large
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17275511
View Documents
Uvex™
8816612
X-Large
124.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17285511
View Documents
Uvex™
8816613
2X-Large
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
17295511
View Documents
Uvex™
8816614
3X-Large
124.00 DKK
Each
Only null left
Estimated Shipment: 17-07-2025
Add to basket
Description
Description
  • Black
  • Material: 100% combed cotton, 145 g/m² Single Jersey Classical T-shirt form of combed cotton and Single Jersey.
  • Neck tape
  • Elastic cuffs on neck
  • Color-stable and easy care
  • Size: XS - 6XL
  • Care instructions: 60°C washable, chlorinate impossible, ironing: II-points, tumble dryer impossible
Specifications
Specifications
Men's
Black
Taped
Reusable
140 g/m²
Cotton
Hooded
Long Sleeve
Multiple Use General Purpose Shirts
Documents
Product Certifications
Product Content Correction
Product Title
Uvex™ Men-T-Shirt 7459-Black 100% Cotton