Learn More
Trifluoroacetic Acid, Thermo Scientific
234.99 DKK - 1275.66 DKK
Specifications
Item Name | Trifluoroacetic Acid (TFA) |
---|---|
Form | Liquid |
Reagent Type | Ion Pairing Reagent |
Formulation | 99.5% TFA |
For Use With | LC/MS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30280970
|
Thermo Scientific
T001160001 |
1 mL |
277.51 DKK
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30280966
|
Thermo Scientific
T001160002 |
2 mL |
417.01 DKK
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30280947
|
Thermo Scientific
T001160005 |
5 mL |
234.99 DKK
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30280960
|
Thermo Scientific
T001160050 |
50 mL |
746.00 DKK
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30280948
|
Thermo Scientific
|
10 x 1 mL |
1275.66 DKK
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
- Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
- Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
Trifluoroacetic Acid (TFA) | |
Ion Pairing Reagent | |
LC/MS |
Liquid | |
99.5% TFA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Trifluoroacetic Acid, Thermo Scientific