missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin lambda-like polypeptide 5, was expressed in the hosts and tags indicated.
1500.00 DKK - 15995.00 DKK
Specifications
Accession Number | NP_001171597.1 |
---|---|
Gene ID (Entrez) | 100423062 |
Name | Immunoglobulin lambda-like polypeptide 5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088611
|
enQuireBio™
QP9714-YE-100UG |
100 μg |
4190.00 DKK
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028621
|
enQuireBio™
QP9714-YE-500UG |
500 μg |
11360.00 DKK
500µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098611
|
enQuireBio™
QP9714-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018621
|
enQuireBio™
QP9714-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038621
|
enQuireBio™
QP9714-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008621
|
enQuireBio™
QP9714-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001171597.1 | |
Immunoglobulin lambda-like polypeptide 5 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
100423062 | |
Research Use Only | |
IGLL5 | |
Recombinant Protein | |
Yeast | |
HGLLRPMVAP QSGDPDPGAS VGSSRSSLRS LWGRLLLQPS PQRADPRCWP RGFWSEPQSL CYVFGTGTKV TVLGQPKANP TVTLFPPSSE ELQANKATLV CLISDFYPGA VTVAWKADGS PVKAGVETTK PSKQSNNKYA ASSYLSLTPE QWKSHRSYSC QVTHEGSTVE KTVAPTECS | |
Greater than 90% as determined by SDS-PAGE. |