Learn More
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin lambda-like polypeptide 5, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9714-ye-50ug
 View more versions of this product
Product Code. 16038621
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_001171597.1
Immunoglobulin lambda-like polypeptide 5 Protein
Research Use Only
IGLL5
Recombinant Protein
Yeast
HGLLRPMVAP QSGDPDPGAS VGSSRSSLRS LWGRLLLQPS PQRADPRCWP RGFWSEPQSL CYVFGTGTKV TVLGQPKANP TVTLFPPSSE ELQANKATLV CLISDFYPGA VTVAWKADGS PVKAGVETTK PSKQSNNKYA ASSYLSLTPE QWKSHRSYSC QVTHEGSTVE KTVAPTECS
Greater than 90% as determined by SDS-PAGE.
100423062
50 ÎĽg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein > 50ÎĽg