missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin lambda-like polypeptide 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9714-ye-10ug
This item is not returnable.
View return policy
Specifications
NP_001171597.1 | |
Immunoglobulin lambda-like polypeptide 5 Protein | |
Research Use Only | |
IGLL5 | |
Recombinant Protein | |
Yeast | |
HGLLRPMVAP QSGDPDPGAS VGSSRSSLRS LWGRLLLQPS PQRADPRCWP RGFWSEPQSL CYVFGTGTKV TVLGQPKANP TVTLFPPSSE ELQANKATLV CLISDFYPGA VTVAWKADGS PVKAGVETTK PSKQSNNKYA ASSYLSLTPE QWKSHRSYSC QVTHEGSTVE KTVAPTECS | |
Greater than 90% as determined by SDS-PAGE. |
100423062 | |
10 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction