Learn More
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin lambda-like polypeptide 5, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9714-ye-50ug
 View more versions of this product
Product Code. 16038621
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
NP_001171597.1
Immunoglobulin lambda-like polypeptide 5 Protein
Research Use Only
IGLL5
Recombinant Protein
Yeast
HGLLRPMVAP QSGDPDPGAS VGSSRSSLRS LWGRLLLQPS PQRADPRCWP RGFWSEPQSL CYVFGTGTKV TVLGQPKANP TVTLFPPSSE ELQANKATLV CLISDFYPGA VTVAWKADGS PVKAGVETTK PSKQSNNKYA ASSYLSLTPE QWKSHRSYSC QVTHEGSTVE KTVAPTECS
Greater than 90% as determined by SDS-PAGE.
100423062
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein > 50μg