Learn More
uvex Phynomic Lite White Safety Gloves
Lightweight gloves are constructed from a combination of polyamide and elastane, with aqua polymer impregnation on the palm and fingertips for increased durability and dexterity.
102.00 DKK - 437.00 DKK
Specifications
Certifications/Compliance | OEKO-TEX™ Standard 100 |
---|---|
Applications | Industrial |
Coating Material | Polymer |
Color | White |
Exterior Finish | Smooth |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17230351
|
uvex
6004105 |
5 |
102.00 DKK
1 pair |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
17240351
|
uvex
6004106 |
6 |
102.00 DKK
1 pair |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
17250351
|
uvex
6004107 |
7 |
437.00 DKK
10 pairs |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
17260351
|
uvex
6004108 |
8 |
437.00 DKK
10 pairs |
|
||||||
17270351
|
uvex
6004109 |
9 |
437.00 DKK
10 pairs |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17280351
|
uvex
6004110 |
10 |
437.00 DKK
10 pairs |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
17290351
|
uvex
6004111 |
11 |
437.00 DKK
10 pairs |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
17200361
|
uvex
6004112 |
12 |
437.00 DKK
10 pairs |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
Description
- Extremely lightweight construction makes these gloves suitable for particularly delicate tasks
- Aqua-polymer impregnation on the palm and fingertips for mechanical abrasion resistance
- Enhances grip capabilities in dry and slightly damp areas
- Porous coating is breathable to reduce perspiration
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
- Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
- 3D Ergo technology ensures a secure fit
- Suitable for use with food handling and processing
Specifications
OEKO-TEX™ Standard 100 | |
Polymer | |
Smooth | |
Yes | |
Mechanical Protection Gloves | |
Driving/Operating Machinery, Painting, and Paper Hanging | |
Knit | |
15 |
Industrial | |
White | |
Ambidextrous | |
Elastane, Polyamide | |
2121X | |
Unisex | |
Phynomic Lite |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Phynomic Lite White Safety Gloves