Learn More
uvex Phynomic All-Round Safety Gloves
Lightweight, dirt-resistant gloves are constructed from a blend of polyamide and elastane, with aqua-polymer foam coating on the palm and fingertips to repel moisture.
447.00 DKK
Specifications
Certifications/Compliance | OEKO-TEX™ Standard 100 |
---|---|
Applications | Industrial |
Coating Material | Aqua-Polymer Foam |
Color | Gray, Black |
Exterior Finish | Smooth |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17210361
|
uvex
6004905 |
5 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17220361
|
uvex
6004906 |
6 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17230361
|
uvex
6004907 |
7 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17240361
|
uvex
6004908 |
8 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17250361
|
uvex
6004909 |
9 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17260361
|
uvex
6004910 |
10 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17270361
|
uvex
6004911 |
11 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17280361
|
uvex
6004912 |
12 |
447.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
Description
- Lightweight construction acts as a second-skin to maintain a high level of feeling
- Aqua-polymer foam coating is moisture repellent and flexible
- Breathable materials help prevent excessive perspiration
- Dirt-resistant exterior is ideal for mechanical tasks
- Enhanced grip in dry and slightly damp applications
- 3D Ergo technology ensures a secure fit
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
- Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
Specifications
OEKO-TEX™ Standard 100 | |
Aqua-Polymer Foam | |
Smooth | |
Yes | |
Mechanical Protection Gloves | |
Assembly, Maintanence, Shipping/Logistics, Precision Work, and Repairs | |
Knit | |
15 |
Industrial | |
Gray, Black | |
Ambidextrous | |
Polyamide, Elastane | |
3131X | |
Unisex | |
Phynomic All-Round |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Phynomic All-Round Safety Gloves