missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ DOB Medium, Powdered Media
Can be used to create SD or SDA by combining with the appropriate amino acid mixture
Brand: MP Biomedicals™ 114025032
3984.15 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Use 26.7g powder per liter
- Contents/liter: 1.7g YNB, 5g Ammonium Sulfate, 20g Dextrose
Yeast cell culture, Gene expression

Specifications
Molecular biology Certified™ | |
Powder | |
YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L | |
1000 g |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ DOB Medium, Powdered Media > 1kg
Spot an opportunity for improvement?Share a Content Correction