missing translation for 'onlineSavingsMsg'
Learn More
Lucitanib, MedChemExpress Product Code.: 16496478

Lucitanib, MedChemExpress

Product Code. p-200101581
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496478

Brand: Medchem Express HY15391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lucitanib (E-3810) is a novel dual inhibitor of VEGFR and FGFR, potently and selectively inhibits VEGFR1, VEGFR2, VEGFR3, FGFR1 and FGFR2 with IC50s of 7 nM, 25 nM, 10 nM, 17.5 nM, and 82.5 nM, respectively.

Lucitanib (E-3810) is a novel dual inhibitor of VEGFR and FGFR, potently and selectively inhibits VEGFR1, VEGFR2, VEGFR3, FGFR1 and FGFR2 with IC50s of 7 nM, 25 nM, 10 nM, 17.5 nM, and 82.5 nM, respectively.

Specifications

Chemical Name or Material Lucitanib
CAS 1058137-23-7
Color White
Purity Grade Notes Research
Molecular Formula C26H25N3O4
Quantity 10 mg
Synonym E-3810
Solubility Information DMSO : ≥ 25 mg/mL (56.37 mM)
SMILES O=C(NC)C1=CC=CC2=C1C=CC(OC3=CC=NC4=CC(OCC5(N)CC5)=C(OC)C=C43)=C2
Molecular Weight (g/mol) 443.49
Formula Weight 443.49
Percent Purity 96.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Lucitanib, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.