missing translation for 'onlineSavingsMsg'
Learn More
KB-R7943 mesylate, MedChemExpress Product Code.: 16476498

KB-R7943 mesylate, MedChemExpress

Product Code. p-200100480
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16476498

Brand: Medchem Express HY15415

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KB-R7943 mesylate is a widely used inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with IC50 of 5.7±2.1 μM. KB-R7943 mesylate induces cancer cell death via activating the JNK pathway and blocking autophagic flux.

KB-R7943 mesylate is a widely used inhibitor of the reverse Na+/Ca2+ exchanger (NCXrev) with IC50 of 5.7±2.1 μM. KB-R7943 mesylate induces cancer cell death via activating the JNK pathway and blocking autophagic flux.

Specifications

Chemical Name or Material KB-R7943 mesylate
CAS 182004-65-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H21N3O6S2
Quantity 10 mg
Solubility Information DMSO : ≥ 27 mg/mL (63.16 mM) ∣H2O : 4.3 mg/mL (10.06 mM; Need warming)
SMILES NC(SCCC1=CC=C(OCC2=CC=C([N+]([O-])=O)C=C2)C=C1)=N.CS(=O)(O)=O
Molecular Weight (g/mol) 427.5
Formula Weight 427.5
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
KB-R7943 mesylate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.