CAS RN 88899-55-2
CAS RN 88899-55-2
IUPAC Name:
(3Z,5E,7R,8S,9S,11E,13E,15S,16R)-16-[(2S,3R,4S)-4-[(2R,4R,5S,6R)-2,4-dihydroxy-5-methyl-6-(propan-2-yl)oxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
Synonyms:
bafilomycin A1 bafilomycin
Molecular Weight (g/mol):
622.84
Molecular Formula:
C35H58O9
InChi Key:
XDHNQDDQEHDUTM-JQWOJBOSSA-N
SMILES:
CO[C@H]1\C=C\C=C(C)\C[C@H](C)[C@H](O)[C@H](C)\C=C(/C)\C=C(OC)\C(=O)O[C@@H]1[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](O)[C@H](C)[C@H](O1)C(C)C
1
–
5
of
5
results
2
Tocris Bioscienceâ„¢ Bafilomycin A1
H+-ATPase (vacuolar) inhibitor
3
Melford Bafilomycin A1
Bafilomycin A1 is a potent and selective inhibitor of the vacuolar-type H+-ATPase (V-ATPase), an enzyme responsible for maintaining acidic pH in cellular compartments such as lysosomes and endosomes.
4
Medchem Express Bafilomycin A1, MedChemExpress
MedChemExpress Bafilomycin A1 (BafA1) is a specific and reversible inhibitor of vacuolar H+-ATPase (V-ATPase) with IC50 values of 4-400 nmol/mg. Bafilomycin A1, a macrolide antibiotic, is also used as an autophagy inhibitor at the late stage. Bafilomycin A1 blocks autophagosome-lysosome fusion and inhibits acidification and protein degradation in lysosomes of cultured cells. Bafilomycin A1 induces apoptosis.

5