CAS RN 2491-36-3
CAS RN 2491-36-3
IUPAC Navn:
2-bromo-1-(2-hydroxyphenyl)ethan-1-one
Synonymer:
2-bromo-1-(2-hydroxyphenyl)ethanone
Molekylvægt (g/mol):
215.05
Molekylformel:
C8H7BrO2
InChi Key:
SGPKEYSZPHMVNI-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1C(=O)CBr
1
–
4
af
4
Resultater
1
2-brom-2'-hydroxyacetophenon, 97 %, Thermo Scientific Chemicals
CAS: 2491-36-3 Molekylær formel: C8H7BrO2 Molekylvægt (g/mol): 215.046 MDL nummer: MFCD01727570 InChI nøgle: SGPKEYSZPHMVNI-UHFFFAOYSA-N PubChem CID: 200671 IUPAC navn: 2-brom-1-(2-hydroxyphenyl)ethanon SMIL: C1=CC=C(C(=C1)C(=O)CBr)O
2
2-Bromo-2’-hydroxyacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Bromo-2’-hydroxyacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-Bromo-2’-hydroxyacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.