accessibility menu, dialog, popup

Test

CAS RN 103222-11-3

CAS RN 103222-11-3

IUPAC Name: 2-amino-N-[10-(4-aminobutyl)-4-{[1-carbamoyl-2-(1H-indol-3-yl)ethyl]carbamoyl}-16-[(4-hydroxyphenyl)methyl]-13-[(1H-indol-3-yl)methyl]-6,9,12,15,18-pentaoxo-7-(propan-2-yl)-1,2-dithia-5,8,11,14,17-pentaazacycloicosan-19-yl]-3-phenylpropanamide
Synonyms: vapreotide
Molecular Weight (g/mol): 1131.38
Molecular Formula: C57H70N12O9S2
InChi Key: SWXOGPJRIDTIRL-UHFFFAOYNA-N
SMILES: CC(C)C1NC(=O)C(CCCCN)NC(=O)C(CC2=CNC3=CC=CC=C23)NC(=O)C(CC2=CC=C(O)C=C2)NC(=O)C(CSSCC(NC1=O)C(=O)NC(CC1=CNC2=CC=CC=C12)C(N)=O)NC(=O)C(N)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)