CAS RN 608512-97-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
6
af
6
Resultater
1
Imidazolo-oxindole PKR inhibitor C16(PKR Inhibitor), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Imidazolo-oxindole PKR inhibitor C16(PKR Inhibitor), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Imidazolo-oxindole PKR inhibitor C16(PKR Inhibitor), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Tocris Bioscience™ C16
PKR inhibitor
5
6