Få mere at vide
Tocris Bioscience™ C16
PKR inhibitor
Brand: Tocris Bioscience™ 5382/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
C16 is a PKR inhibitor (IC50 = 210 nM). Rescues PKR-dependent translation block in vitro. Decreases Aβ42-induced inflammatory cytokine release and apoptosis in neuronal cultures. Prevents neuroinflammation and neuronal loss in an acute excitotoxic rat model. Brain penetrant.Tekniske data
| C16 | |
| 6,8-Dihydro-8-(1H-imidazol-5-ylmethylene)-7H-pyrrolo[2,3-g]benzothiazol-7-one | |
| Soluble to 50mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 608512-97-6 | |
| 10 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use