Få mere at vide
Tocris Bioscience™ C16
Beskrivelse
C16 is a PKR inhibitor (IC50 = 210 nM). Rescues PKR-dependent translation block in vitro. Decreases Aβ42-induced inflammatory cytokine release and apoptosis in neuronal cultures. Prevents neuroinflammation and neuronal loss in an acute excitotoxic rat model. Brain penetrant.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | C16 |
| CAS | 608512-97-6 |
| Synonym | 6,8-Dihydro-8-(1H-imidazol-5-ylmethylene)-7H-pyrrolo[2,3-g]benzothiazol-7-one |
| Mængde | 10 mg |
| Opløselighed | Soluble to 50mM in DMSO and to 20mM in ethanol |
| Molekylær formel | C13H8N4OS |
| Formel vægt | 268.29 |
| Fysisk form | Solid |
| Produkttype | Bioactive Small Molecule |
| Renhed | >98% |
Research Use Only. Not For Human or Veterinary Use