CAS RN 56341-41-4
IUPAC Navn:
5-fluoro-2,3-dihydro-1H-indol-2-one
Synonymer:
5-fluoro-1,3-dihydroindol-2-one
Molekylvægt (g/mol):
151.14
Molekylformel:
C8H6FNO
InChi Key:
DDIIYGHHUMKDGI-UHFFFAOYSA-N
SMILES:
FC1=CC=C2NC(=O)CC2=C1
1
–
4
af
4
Resultater
2
5-Fluorooxindole (5-Fluoro-1,3-dihydro-2H-indol-2-one), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
5-Fluoro-2-oxindole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.