CAS RN 503468-95-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
NU 7441, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
NU 7441, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4
KU-57788, MedChemExpress
MedChemExpress KU-57788 (NU7441) is a highly potent and selective DNA-PK inhibitor with an IC50 of 14 nM. KU-57788 is an NHEJ pathway inhibitor. KU-57788 also inhibits PI3K and mTOR with IC50s of 5.0 and 1.7 μM, respectively.