missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies NU7441

Artikelnummer. 17160322
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17160322 10 mg
17150322 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17160322 Leverandør STEMCELL Technologies Leverandørnr. 74084

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

NHEJ pathway inhibitor; Inhibits DNA-dependent protein kinase (DNA-PK)

  • NU7441 is an inhibitor of DNA-dependent protein kinase (DNA-PK), an enzyme involved in the non-homologous end joining (NHEJ) DNA repair pathway
  • It is highly selective for DNA-PK, with an IC50 of 14 nM (Leahy et al.)
  • Reduces the frequency of NHEJ and increases the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Robert et al.)
  • Sensitizes human cancer cell lines to DNA double-strand-break-inducing therapies (chemo- or radio-therapy) by inhibiting DNA-PK activity and delaying the repair of double-strand breaks (Ciszewski et al.; Shaheen et al.; Yang et al.; Zhao et al.)

Tekniske data

CAS 503468-95-9
Synonym KU57788
Mængde 10 mg
Molekylær formel C25H19NO3S
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.