CAS RN 2303-17-5
IUPAC Navn:
N,N-bis(propan-2-yl)[(2,3,3-trichloroprop-2-en-1-yl)sulfanyl]formamide
Synonymer:
triallate
Molekylvægt (g/mol):
304.65
Molekylformel:
C10H16Cl3NOS
InChi Key:
MWBPRDONLNQCFV-UHFFFAOYSA-N
SMILES:
CC(C)N(C(C)C)C(=O)SCC(Cl)=C(Cl)Cl
1
–
7
af
7
Resultater
2
Triallate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Triallate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Triallate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Tri-allate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality