Få mere at vide
Tri-Allate, SPEX CertiPrep™
Artikelnummer. 12959384 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12959384 1 mL 1ml
1 options
Artikelnummer. 12959384 Leverandør SPEX Certiprep Leverandørnr. S3550
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Tri-allate2303-17-51000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale Tri-allate
Emballage 2mL amber prescored glass ampule
CAS 2303-17-5, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C10H16Cl3NOS
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle MWBPRDONLNQCFV-UHFFFAOYSA-N
SMIL CC(C)N(C(C)C)C(=O)SCC(=C(Cl)Cl)Cl
IUPAC navn S-(2,3,3-trichloroprop-2-enyl) N,N-di(propan-2-yl)carbamothioate
Opløsningsmiddel eller Matrix Methanol
Molekylvægt (g/mol) 304.654
PubChem CID 5543
Mængde 1 mL
ChEBI CHEBI:81978
Vis mere Vis mindre
Product intended for laboratory use only