CAS RN 1798871-30-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Centrinone, TRC
CAS: 1798871-30-3 Molekylær formel: C26H25F2N7O6S2 Molekylvægt (g/mol): 633.65 Synonym: 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(4-morpholinyl)-4-pyrimidinamine IUPAC navn: 2-[2-fluoro-4-[(2-fluoro-3-nitrophenyl)methylsulfonyl]phenyl]sulfanyl-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-morpholin-4-ylpyrimidin-4-amine SMIL: FC1=C(C=CC=C1CS(C2=CC=C(C(F)=C2)SC3=NC(N4CCOCC4)=C(C(NC5=NNC(C)=C5)=N3)OC)(=O)=O)[N+]([O-])=O
2
Tocris Bioscience™ Centrinone
High affinity and selective PLK4 inhibitor
3
Centrinone, MedChemExpress
MedChemExpress Centrinone (LCR-263) is a selective and reversible inhibitor of polo-like kinase 4 (PLK4) with a Ki of 0.16 nM.
4