Få mere at vide
Tocris Bioscience™ Centrinone
High affinity and selective PLK4 inhibitor
Brand: Tocris Bioscience™ 5687/10
Beskrivelse
High affinity and selective PLK4 inhibitor (Ki = 0.16 nM). Exhibits >1000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner. Decreases expression of pluripotency markers and induces differentiation in pluripotent stem cells.
Centrinone is a high affinity and selective PLK4 inhibitor (Ki = 0.16 nM). Exhibits >1000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner. Decreases expression of pluripotency markers and induces differentiation in pluripotent stem cells.Tekniske data
| 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(4-morpholinyl)-4-pyrimidinamine | |
| 1798871-30-3 | |
| 10 mg |