CAS RN 16727-10-9
IUPAC Navn:
N,N-dimethylacetamide
Synonymer:
dimethylacetamide hallucinogen N,N'-dimethylacetamide
Molekylvægt (g/mol):
87.12
Molekylformel:
C4H9NO
InChi Key:
FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES:
CN(C)C(C)=O
1
–
5
af
5
Resultater
1
N,N-Dimethylacetamide-d9, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N-Dimethylacetamide-d9, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N,N-Dimethylacetamide-d9, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
4
N,N-Dimethylacetamide-d9, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.