Få mere at vide
N,N-Dimethylacetamide-d9, TRC
Artikelnummer. 30368881
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30368881 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368881 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47115850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Dimethylacetamide (D9)
CAS 16727-10-9
Molekylær formel C4D9NO
Mængde 50 mg
Synonym Acetamide-2,2,2-d3, N,N-di(methyl-d3)- (8CI, 9CI, ACI), N,N-Di(methyl-d3)acetamide-2,2,2-d3 (ACI), 2,2,2-Trideuterio-N,N-bis(trideuteriomethyl)acetamide, N,N-Dimethylacetamide-d9, N,N-Dimethylacetamide-D9
InChI formel InChI=1S/C4H9NO/c1-4(6)5(2)3/h1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])N(C(=O)C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2,2,2-trideuterio-N,N-bis(trideuteriomethyl)acetamide
Molekylvægt (g/mol) 96.176
Formel vægt 96.1249