Learn More
RETSCH Zirconium Oxide Grinding Jars
Spare Grinding Jar
9255.00 DKK - 59440.00 DKK
Product Code | Brand | Capacity (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
15380964
|
RETSCH
01.462.0307 |
125 mL | For EMAX |
59440.00 DKK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
14841631
|
RETSCH
01.462.0336 |
25 mL | For CryoMill |
17580.00 DKK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15390964
|
RETSCH
01.462.0312 |
50 mL | For EMAX |
48305.00 DKK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
10623011
|
RETSCH
01.462.0195 |
25 mL | For MM 200 Mixer Mill |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10636141
|
RETSCH
01.462.0194 |
10 mL | For MM 200 Mixer Mill |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Accessory
Specifications
Grinding Jar | |
Zirconium Oxide |