Learn More
BRAND™ X-Shape PTFE Magnetic Stirring Bars
An excellent option for liquid agitation in the laboratory.
433.00 DKK - 932.88 DKK
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||||
10683012
|
BRAND™
137630 |
10 mm | 5mm |
433.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10598401
|
BRAND™
137632 |
20 mm | 9 mm |
513.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10508221
|
BRAND™
137634 |
25 mm | 10mm |
595.00 DKK
Pack of 10 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
10749011
|
BRAND™
137636 |
30 mm | 12mm |
820.00 DKK
Pack of 10 |
Estimated Shipment: 24-06-2025
Log in to see stock. |
|||||
11814100
|
BRAND™
137638 |
38 mm | 15mm |
932.88 DKK
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Steady spinning position
- Excellent stirring action
- Provided in packs of 10
Specifications
PTFE | |
10/Pk. | |
Cross |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ X-Shape PTFE Magnetic Stirring Bars