Learn More
Tempshield™ Waterproof Cryo-Grip™ Gloves
Waterproof Cryo-Grip™ Gloves give superior dexterity and hand protection when working with liquid nitrogen. The multi-layered construction allows for a maximum level of thermal protection and the thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
3140.00 DKK - 3345.00 DKK
Specifications
Certifications/Compliance (Europe) | CE Cat. III, EN 511, EN 388, EN 420 |
---|---|
Color | Pink |
Product Type | Cryogenic Gloves |
Temperature (Metric) Max | -196°C |
For Use With (Application) | Handling of smaller objects or samples stored or hardened in cryogenic liquids, dispensing/transferring cryogenic liquids, any open cryogenic containers with the chance of exposure to liquids or splashes |
Product Code | Brand | Style | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Style | Size | Price | Quantity & Availability | |||||
16022252
|
Tempshield™
P-CGWRSWP |
Wrist | Small |
3140.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16002242
|
Tempshield™
P-CGMASWP |
Mid-Arm | Small |
3345.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16012252
|
Tempshield™
P-CGWRMWP |
Wrist | Medium |
3140.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16092232
|
Tempshield™
P-CGMAMWP |
Mid-Arm | Medium |
3345.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16082232
|
Tempshield™
P-CGMALWP |
Mid-Arm | Large |
3345.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
Description
- Grip palm for improved dexterity handling small objects
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- High level of thermal protection when exposed to cryogenic fluids
- For ultra-cold applications down to -196°C
- CE Category III: EN 511, EN 388, EN 420
Specifications
CE Cat. III, EN 511, EN 388, EN 420 | |
Cryogenic Gloves | |
Handling of smaller objects or samples stored or hardened in cryogenic liquids, dispensing/transferring cryogenic liquids, any open cryogenic containers with the chance of exposure to liquids or splashes |
Pink | |
-196°C | |
Tempshield™ Cyro-Protection™ |