Learn More
Tempshield™ Waterproof Cryo-Grip™ Gloves
Waterproof Cryo-Grip™ Gloves - Superior dexterity and hand protection when working with liquid nitrogen. The multi-layered construction allows for a maximum level of thermal protection and the thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
3140.00 DKK - 4980.00 DKK
Specifications
Certifications/Compliance (Europe) | CE Cat. III, EN 511, EN 388, EN 420 |
---|---|
Color | Blue |
Product Type | Cryogenic Gloves |
Temperature (Metric) Max | -196°C |
For Use With (Application) | Handling of smaller objects or samples stored or hardened in cryogenic liquids, dispensing/transferring cryogenic liquids, any open cryogenic containers with the chance of exposure to liquids or splashes |
Product Code | Brand | Size | Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Style | Price | Quantity & Availability | |||||
16072222
|
Tempshield™
EBSWP |
Small | Elbow |
3500.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16022222
|
Tempshield™
CGWRMWP |
Medium | Wrist |
3140.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16002212
|
Tempshield™
CGMAMWP |
Medium | Mid-Arm |
3345.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
16042202
|
Tempshield™
CGEBMWP |
Medium | Elbow |
4010.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 13-11-2025
Log in to see stock. |
|||||
16012222
|
Tempshield™
CGWRLWP |
Large | Wrist |
3140.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16032202
|
Tempshield™
CGEBLWP |
Large | Elbow |
4285.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
16092202
|
Tempshield™
CGMALWP |
Large | Mid-Arm |
3345.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
16052212
|
Tempshield™
CGSHLWP |
Large | Shoulder |
4980.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
16022212
|
Tempshield™
CGMAXLWP |
X-Large | Mid-Arm |
3345.00 DKK
1 pair Qty Price Break Available |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
Description
- Grip palm for improved dexterity handling small objects
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- High level of thermal protection when exposed to cryogenic fluids
- For ultra-cold applications down to -196°C (-320°F)
- Maximum level of thermal and splash protection, flexibility, and dexterity
- CE Category III: EN 511, EN 388, EN 420
- Applications: handling of smaller objects or samples that have been stored or hardened in cryogenic liquids, dispensing/transferring cryogenic liquids, any open cryogenic containers with the chance of exposure to liquids or splashes.
Specifications
CE Cat. III, EN 511, EN 388, EN 420 | |
Cryogenic Gloves | |
Handling of smaller objects or samples stored or hardened in cryogenic liquids, dispensing/transferring cryogenic liquids, any open cryogenic containers with the chance of exposure to liquids or splashes |
Blue | |
-196°C | |
Tempshield™ Cyro-Protection™ |