Learn More
OHAUS™ Uni Block
Hot plates and stirrers accessory
Brand: OHAUS™ 30500583
2454.66 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Holds test tubes or vials
Specifications
17 mm Vials | |
Uni Block | |
Metal |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
OHAUS™ Uni Block > 17 mm Vials
Spot an opportunity for improvement?Share a Content Correction