Learn More
Metro™ Super Erecta™ Wire Shelf, Chrome Finish
Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in chrome finish allow adjustable configurations when used with Metro Super Erecta shelving systems
640.00 DKK - 2155.00 DKK
Specifications
For Use With (Equipment) | Metro™ Super Erecta™ shelving system |
---|---|
Includes | Plastic sleeves and wedges |
Material | Chrome |
Product Code | Brand | Load Bearing Capacity (English) | Load Bearing Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity (English) | Load Bearing Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
15976591
|
Metro™
1836NC |
400 lb. | 363 kg | 36 x 18 in. (91.4 x 45.7 cm) |
650.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15946601
|
Metro™
1848NC |
400 lb. | 363 kg | 48 x 18 in. (121.9 x 45.7 cm) |
785.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15906611
|
Metro™
1860NC |
600 lb. | 272 kg | 60 x 18 in. (152.4 x 45.7 cm) |
905.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15976681
|
Metro™
3060NC |
400 lb. | 182 kg | 60 x 30 in. (152.4 x 76 cm) |
1550.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15926701
|
Metro™
3648NC |
600 lb. | 272 kg | 48 x 36 in. (121.9 x 91 cm) |
1615.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15906661
|
Metro™
2436NC |
400 lb. | 363 kg | 36 x 24 in. (91.4 x 61 cm) |
835.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15966571
|
Metro™
1442NC |
800 lb. | 363 kg | 42 x 14 in. (106.6 x 35.5 cm) |
710.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15926581
|
Metro™
1460NC |
600 lb. | 272 kg | 60 x 14 in. (152.4 x 35.5 cm) |
825.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15966611
|
Metro™
2124NC |
800 lb. | 363 kg | 24 x 21 in. (61 x 53 cm) |
750.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15996581
|
Metro™
1824NC |
800 lb. | 363 kg | 24 x 18 in. (61 x 45.7 cm) |
640.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15916621
|
Metro™
2130NC |
800 lb. | 363 kg | 30 x 21 in. (76 x 53 cm) |
780.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15966621
|
Metro™
2136NC |
800 lb. | 363 kg | 36 x 21 in. (91.4 x 53 cm) |
780.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15956631
|
Metro™
2148NC |
800 lb. | 363 kg | 48 x 21 in. (122 x 53 cm) |
895.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15906631
|
Metro™
2142NC |
800 lb. | 363 kg | 42 x 21 in. (106.6 x 53 cm) |
895.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15976641
|
Metro™
2172NC |
600 lb. | 272 kg | 72 x 21 in. (183 x 53 cm) |
1270.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15936641
|
Metro™
2160NC |
600 lb. | 272 kg | 60 x 21 in. (152.4 x 53 cm) |
1070.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15956651
|
Metro™
2430NC |
800 lb. | 363 kg | 30 x 24 in. (76 x 61 cm) |
835.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15956661
|
Metro™
2442NC |
800 lb. | 363 kg | 42 x 24 in. (106.6 x 61 cm) |
965.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15906701
|
Metro™
3636NC |
800 lb. | 363 kg | 36 x 36 in. (91 x 91 cm) |
1390.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15906691
|
Metro™
3072NC |
600 lb. | 272 kg | 72 x 30 in. (183 x 76 cm) |
1850.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15966701
|
Metro™
3672NC |
600 lb. | 272 kg | 72 x 36 in. (183 x 91 cm) |
2155.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15936681
|
Metro™
3036NC |
800 lb. | 363 kg | 36 x 30 in. (91.4 x 76 cm) |
1220.00 DKK
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
15956671
|
Metro™
2460NC |
21 lb. | 9.5 kg | 60 x 24 in. (152.4 x 61 cm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Real nickel-chrome finish for dry, low humidity environment
- Protective lacquer coated
Specifications
Metro™ Super Erecta™ shelving system | |
Plastic sleeves and wedges | |
Chrome |