missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., anhydrous, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
40 kg
6 x 1 kg
Unit Size:
1kg
40kg
500g
5kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 500 g |
Linear Formula | CH3COONa |
UN Number | NONH for all modes of transport |
Beilstein | 3595639 |
Flash Point | >250°C (>482°F) |
Formula Weight | 82.03g/mol |
Grade | ACS/puriss. p.a. |
Ignition Point | 1112°F |
Chemical Name or Material | Sodium Acetate, Reag. Ph. Eur. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., anhydrous, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction