Learn More
uvex Rubiflex S NB27S Chemical Protection Gloves
Gloves are fully coated with NBR for chemical resistance, and feature a high-quality cotton interlock liner that provides heat insulation for increased mechanical protection.
1115.00 DKK
Specifications
Applications | Laboratory |
---|---|
Coating Material | NBR |
Color | Green |
Latex Free | Yes |
Material | Nitrile |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17230242
|
uvex
9893002 |
8 |
1115.00 DKK
10 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17270242
|
uvex
9893059 |
9 |
1115.00 DKK
10 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17240252
|
uvex
9893730 |
10 |
1115.00 DKK
10 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17240242
|
uvex
9893005 |
11 |
1115.00 DKK
10 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
Description
- NBR-coated safety gloves with reinforced cotton interlock supporting material
- Thick, reinforced liner provides insulation for thermal protection
- Resistant to a variety of chemicals, acids, alkalis, mineral oils, and solvents
- Suitable for applications that require protection against chemicals and various mechanical risks
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
Specifications
Laboratory | |
Green | |
Nitrile | |
2121X | |
JKNOPT | |
Unisex | |
Cotton | |
27 cm |
NBR | |
Yes | |
Chemical-resistant Gloves | |
Type A | |
Automotive, Chemical Handling, Food Processing, Machine Operation, Metal Work, and Sandblasting | |
Gauntlet | |
Rubiflex S NB27S |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Rubiflex S NB27S Chemical Protection Gloves