Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
905.00 DKK - 12785.00 DKK
Product Code | Brand | Standard Taper Joint No. | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Includes | Price | Quantity & Availability | ||||
10203180
|
SVL™
RF13 |
13 | Female joint |
905.00 DKK
Pack of 5 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
10228064
|
SVL™
RF19 |
19 | Female joint |
1110.00 DKK
Pack of 5 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
10422395
|
SVL™
RM19 |
19 | Male joint |
1110.00 DKK
Pack of 5 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
12501406
|
SVL™
RF29 |
29 | Female joint |
1655.00 DKK
Pack of 5 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
12079042
|
SVL™
RM29 |
29 | Male joint |
1655.00 DKK
Pack of 5 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
15567950
|
SVL™
RF35 |
35 | Female joint, PTFE covered O-ring, spring loaded Metal clip |
11735.00 DKK
Pack of 30 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
15577950
|
SVL™
RF41 |
41 | Female joint, PTFE covered O-ring, spring loaded Metal clip |
12785.00 DKK
Pack of 30 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
15507960
|
SVL™
RM41 |
41 | Male joint, PTFE covered O-ring, spring loaded Metal clip |
12785.00 DKK
Pack of 30 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
15587950
|
SVL™
RF64 |
64 | Female joint |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15517960
|
SVL™
RM64 |
64 | Male joint |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200°C
- System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
- Joint can be retained together with spring loaded metal clip
- Component parts RM, RF, RO and RC should be ordered separately
Specifications
Glass, PTFE | |
Spherical joint |