Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Brand:  SVL™ RF41
12145.75 DKK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 15577950
  • 12785.00 DKK / Pack of 30
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200°C
  • System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
  • Joint can be retained together with spring loaded metal clip
  • Component parts RM, RF, RO and RC should be ordered separately
Specifications
Specifications
Female joint, PTFE covered O-ring, spring loaded Metal clip
Glass, PTFE
41