missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Gamma-hemolysin component B Protein
A DNA sequence encoding the Staphylococcus aureus (strain N315) Gamma-hemolysin component B, was expressed in the hosts and tags indicated.
2300.00 DKK - 22245.00 DKK
Specifications
Accession Number | WP_000783428.1 |
---|---|
Gene ID (Entrez) | 31215165 |
Name | S. aureus Gamma-hemolysin component B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068110
|
enQuireBio™
QP7309-EC-100UG |
100 μg |
6415.00 DKK
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088110
|
enQuireBio™
QP7309-EC-1MG |
1 mg |
22245.00 DKK
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078110
|
enQuireBio™
QP7309-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098110
|
enQuireBio™
QP7309-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008120
|
enQuireBio™
QP7309-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018120
|
enQuireBio™
QP7309-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000783428.1 | |
S. aureus Gamma-hemolysin component B Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
31215165 | |
Research Use Only | |
hlgB | |
Recombinant Protein | |
E. coli | |
AEGKITPVSV KKVDDKVTLY KTTATADSDK FKISQILTFN FIKDKSYDKD TLVLKATGNI NSGFVKPNPN DYDFSKLYWG AKYNVSISSQ SNDSVNVVDY APKNQNEEFQ VQNTLGYTFG GDISISNGLS GGLNGNTAFS ETINYKQESY RTTLSRNTNY KNVGWGVEAH KIMNNGWGPY GRDSFHPTYG NELFLAGRQS SAYAGQNFIA QHQMPLLSRS NFNPEFLSVL SHRQDGAKKS KITVTYQREM DLYQIRWNGF YWAGANYKNF KTRTFKSTYE IDWENHKVKL LDTKETENNK | |
Greater than 80% as determined by SDS-PAGE. |