missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Gamma-hemolysin component B Protein
A DNA sequence encoding the Staphylococcus aureus (strain N315) Gamma-hemolysin component B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7309-ec-100ug
This item is not returnable.
View return policy
Specifications
WP_000783428.1 | |
S. aureus Gamma-hemolysin component B Protein | |
Research Use Only | |
hlgB | |
Recombinant Protein | |
E. coli | |
AEGKITPVSV KKVDDKVTLY KTTATADSDK FKISQILTFN FIKDKSYDKD TLVLKATGNI NSGFVKPNPN DYDFSKLYWG AKYNVSISSQ SNDSVNVVDY APKNQNEEFQ VQNTLGYTFG GDISISNGLS GGLNGNTAFS ETINYKQESY RTTLSRNTNY KNVGWGVEAH KIMNNGWGPY GRDSFHPTYG NELFLAGRQS SAYAGQNFIA QHQMPLLSRS NFNPEFLSVL SHRQDGAKKS KITVTYQREM DLYQIRWNGF YWAGANYKNF KTRTFKSTYE IDWENHKVKL LDTKETENNK | |
Greater than 80% as determined by SDS-PAGE. |
31215165 | |
100 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. aureus Gamma-hemolysin component B Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction