Learn More
enQuireBio™ Recombinant S. aureus Gamma-hemolysin component B Protein
A DNA sequence encoding the Staphylococcus aureus (strain N315) Gamma-hemolysin component B, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7309-ec-50ug
 View more versions of this product
Product Code. 16018120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
WP_000783428.1
S. aureus Gamma-hemolysin component B Protein
Research Use Only
hlgB
Recombinant Protein
E. coli
AEGKITPVSV KKVDDKVTLY KTTATADSDK FKISQILTFN FIKDKSYDKD TLVLKATGNI NSGFVKPNPN DYDFSKLYWG AKYNVSISSQ SNDSVNVVDY APKNQNEEFQ VQNTLGYTFG GDISISNGLS GGLNGNTAFS ETINYKQESY RTTLSRNTNY KNVGWGVEAH KIMNNGWGPY GRDSFHPTYG NELFLAGRQS SAYAGQNFIA QHQMPLLSRS NFNPEFLSVL SHRQDGAKKS KITVTYQREM DLYQIRWNGF YWAGANYKNF KTRTFKSTYE IDWENHKVKL LDTKETENNK
Greater than 80% as determined by SDS-PAGE.
31215165
50 μg
Not determined.
Full Length
S. aureus
His
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant S. aureus Gamma-hemolysin component B Protein > 50μg