Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 variant 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 variant 2, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_003341.1 |
---|---|
Gene ID (Entrez) | 7336 |
Name | Ubiquitin-conjugating enzyme E2 variant 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16066380
|
enQuireBio™
QP6864-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086380
|
enQuireBio™
QP6864-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076380
|
enQuireBio™
QP6864-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096380
|
enQuireBio™
QP6864-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006390
|
enQuireBio™
QP6864-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016390
|
enQuireBio™
QP6864-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003341.1 | |
Ubiquitin-conjugating enzyme E2 variant 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7336 | |
Research Use Only | |
UBE2V2 | |
Recombinant Protein | |
E. coli | |
AVSTGVKVPR NFRLLEELEE GQKGVGDGTV SWGLEDDEDM TLTRWTGMII GPPRTNYENR IYSLKVECGP KYPEAPPSVR FVTKINMNGI NNSSGMVDAR SIPVLAKWQN SYSIKVVLQE LRRLMMSKEN MKLPQPPEGQ TYNN | |
Greater than 90% as determined by SDS-PAGE. |