Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 variant 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 variant 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6864-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_003341.1 | |
Ubiquitin-conjugating enzyme E2 variant 2 Protein | |
Research Use Only | |
UBE2V2 | |
Recombinant Protein | |
E. coli | |
AVSTGVKVPR NFRLLEELEE GQKGVGDGTV SWGLEDDEDM TLTRWTGMII GPPRTNYENR IYSLKVECGP KYPEAPPSVR FVTKINMNGI NNSSGMVDAR SIPVLAKWQN SYSIKVVLQE LRRLMMSKEN MKLPQPPEGQ TYNN | |
Greater than 90% as determined by SDS-PAGE. |
7336 | |
50 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 variant 2 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction