Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 variant 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 variant 2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6864-ec-50ug
 View more versions of this product
Product Code. 16016390
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
NP_003341.1
Ubiquitin-conjugating enzyme E2 variant 2 Protein
Research Use Only
UBE2V2
Recombinant Protein
E. coli
AVSTGVKVPR NFRLLEELEE GQKGVGDGTV SWGLEDDEDM TLTRWTGMII GPPRTNYENR IYSLKVECGP KYPEAPPSVR FVTKINMNGI NNSSGMVDAR SIPVLAKWQN SYSIKVVLQE LRRLMMSKEN MKLPQPPEGQ TYNN
Greater than 90% as determined by SDS-PAGE.
7336
50 ÎĽg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 variant 2 Protein > 50ÎĽg