missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-3, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_003204.2 |
---|---|
Gene ID (Entrez) | 7004 |
Name | Transcriptional enhancer factor TEF-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079340
|
enQuireBio™
QP7553-ec-10ug |
10 ÎĽg | N/A | ||||||
16019350
|
enQuireBio™
QP7553-ec-50ug |
50 ÎĽg | N/A | ||||||
16069340
|
enQuireBio™
QP7553-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16099340
|
enQuireBio™
QP7553-ec-200ug |
200 ÎĽg | N/A | ||||||
16009350
|
enQuireBio™
QP7553-ec-500ug |
500 ÎĽg | N/A | ||||||
16089340
|
enQuireBio™
QP7553-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_003204.2 | |
Transcriptional enhancer factor TEF-3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7004 | |
Research Use Only | |
TEAD4 | |
Recombinant Protein | |
E. coli | |
MYGRNELIAR YIKLRTGKTR TRKQVSSHIQ VLARRKAREI QAKLKDQAAK DKALQSMAAM SSAQIISATA FHSSMALARG PGRPAVSGFW QGALPGQAGT SHDVKPFSQQ TYAVQPPLPL PGFESPAGPA PSPSAPPAPP WQGRSVASSK LWMLEFSAFL EQQQDPDTYN KHLFVHIGQS SPSYSDPYLE AVDIRQIYDK FPEKKGGLKD LFERGPSNAF FLVKFWADLN TNIEDEGSSF YGVSSQYESP ENMIITCSTK VCSFGKQVVE KVETEYARYE NGHYSYRIHR SPLCEYMINF IHKLKHLPEK YMMNSVLENF TILQVVTNRD TQETLLCIAY VFEVSASEHG AQHHIYRLVK E | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-3 Protein