Learn More
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-3, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7553-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16089340
  • 15565.00 DKK / 1mg
Estimated Shipment: 04-07-2024
to see stock.
Specifications
NP_003204.2
Transcriptional enhancer factor TEF-3 Protein
Research Use Only
TEAD4
Recombinant Protein
E. coli
MYGRNELIAR YIKLRTGKTR TRKQVSSHIQ VLARRKAREI QAKLKDQAAK DKALQSMAAM SSAQIISATA FHSSMALARG PGRPAVSGFW QGALPGQAGT SHDVKPFSQQ TYAVQPPLPL PGFESPAGPA PSPSAPPAPP WQGRSVASSK LWMLEFSAFL EQQQDPDTYN KHLFVHIGQS SPSYSDPYLE AVDIRQIYDK FPEKKGGLKD LFERGPSNAF FLVKFWADLN TNIEDEGSSF YGVSSQYESP ENMIITCSTK VCSFGKQVVE KVETEYARYE NGHYSYRIHR SPLCEYMINF IHKLKHLPEK YMMNSVLENF TILQVVTNRD TQETLLCIAY VFEVSASEHG AQHHIYRLVK E
Greater than 80% as determined by SDS-PAGE.
7004
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol