missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7553-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_003204.2 | |
Transcriptional enhancer factor TEF-3 Protein | |
Research Use Only | |
TEAD4 | |
Recombinant Protein | |
E. coli | |
MYGRNELIAR YIKLRTGKTR TRKQVSSHIQ VLARRKAREI QAKLKDQAAK DKALQSMAAM SSAQIISATA FHSSMALARG PGRPAVSGFW QGALPGQAGT SHDVKPFSQQ TYAVQPPLPL PGFESPAGPA PSPSAPPAPP WQGRSVASSK LWMLEFSAFL EQQQDPDTYN KHLFVHIGQS SPSYSDPYLE AVDIRQIYDK FPEKKGGLKD LFERGPSNAF FLVKFWADLN TNIEDEGSSF YGVSSQYESP ENMIITCSTK VCSFGKQVVE KVETEYARYE NGHYSYRIHR SPLCEYMINF IHKLKHLPEK YMMNSVLENF TILQVVTNRD TQETLLCIAY VFEVSASEHG AQHHIYRLVK E | |
Greater than 80% as determined by SDS-PAGE. |
7004 | |
200 ÎĽg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-3 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction